Platinum wire, 0.508mm (0.02in) dia, For ISA Type R or S Standard Grade Thermocouple
Platinum wire, 0.508mm (0.02in) dia, For ISA Type R or S Standard Grade Thermocouple
Platinum wire, 0.508mm (0.02in) dia, For ISA Type R or S Standard Grade Thermocouple
Thermo Scientific Chemicals

Platinum wire, 0.508mm (0.02in) dia, For ISA Type R or S Standard Grade Thermocouple

Form: approx. 4.34g/m Negative leg Type R or S | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 014361.BU
also known as 014361-BU
Price (USD)
349.00
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
349.00
Each
Specifications
AppearanceSilvery white wire
CAS6-4-7440
ChEBICHEBI:33400
Chemical Name or MaterialPlatinum
ColorWhite
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only