Potassium, plasma standard solution, Specpure™ K 10,000μg/mL
Potassium, plasma standard solution, Specpure™ K 10,000μg/mL
Potassium, plasma standard solution, Specpure™ K 10,000μg/mL
Thermo Scientific Chemicals

Potassium, plasma standard solution, Specpure™ K 10,000μg/mL

CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014379.AD
also known as 014379-AD
Price (USD)
86.10
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
86.10
Each
Specifications
CAS7757-79-1
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:63043
Chemical Name or MaterialPotassium
View more
Potassium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only