Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL
Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL
Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL
Thermo Scientific Chemicals

Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014381.AE
also known as 014381-AE
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Specifications
Chemical Name or MaterialSelenium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only