Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL
Thermo Scientific Chemicals

Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014386.AD
also known as 014386-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialZirconium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
EINECS Number231-595-7
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
View more
Zirconium plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only