Cobalt, plasma standard solution, Specpure™ Co 10,000μg/mL
Cobalt, plasma standard solution, Specpure™ Co 10,000μg/mL
Cobalt, plasma standard solution, Specpure™ Co 10,000μg/mL
Thermo Scientific Chemicals

Cobalt, plasma standard solution, Specpure™ Co 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014390.AD
also known as 014390-AD
Price (USD)
73.65
Online Exclusive
82.10
Save 8.45 (10%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
82.10
Save 8.45 (10%)
Each
Specifications
Chemical Name or MaterialCobalt plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H317-H334-H335-H350
Health Hazard 2GHS H Statement
H334-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only