Gold, plasma standard solution, Specpure™ Au 10,000μg/mL
Gold, plasma standard solution, Specpure™ Au 10,000μg/mL
Gold, plasma standard solution, Specpure™ Au 10,000μg/mL
Thermo Scientific Chemicals

Gold, plasma standard solution, Specpure™ Au 10,000μg/mL

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014392.AD
also known as 014392-AD
Price (USD)
328.00
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
328.00
Each
Specifications
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold
DOT InformationHazard Class: 8, Packing Group: II
EINECS Number231-595-7
View more
Gold, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only