Platinum, plasma standard solution, Specpure™ Pt 10,000μg/mL
Platinum, plasma standard solution, Specpure™ Pt 10,000μg/mL
Platinum, plasma standard solution, Specpure™ Pt 10,000μg/mL
Thermo Scientific Chemicals

Platinum, plasma standard solution, Specpure™ Pt 10,000μg/mL

CAS: 7440-6-6 | Pt in 20% HCl
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014397.AE
also known as 014397-AE
Price (USD)
663.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
663.00
Each
Specifications
CAS6-6-7440
Chemical Name or MaterialPlatinum
DOT InformationHazard Class: 8, Packing Group: II
DensityApprox. 1g/cm3
EINECS Number231-595-7
View more
Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only