Specifications
Chemical Name or MaterialCalcium plasma standard solution
DOT InformationHazard Class: 8; Packaging Group: II
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more