Specifications
Chemical Name or MaterialChromium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
EINECS Number231-595-7
Health Hazard 1H290-H314-H335-H351
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more