Palladium, plasma standard solution, Specpure™ Pd 10,000μg/mL
Palladium, plasma standard solution, Specpure™ Pd 10,000μg/mL
Palladium, plasma standard solution, Specpure™ Pd 10,000μg/mL
Thermo Scientific Chemicals

Palladium, plasma standard solution, Specpure™ Pd 10,000μg/mL

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
Catalog number 014432.AD
also known as 014432-AD
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Specifications
CAS5-3-7440
ChEBICHEBI:33363
Chemical Name or MaterialPalladium
DOT InformationHazard Class: 8, Packing Group: II
EINECS Number231-595-7
View more
Palladium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only