Sulfur, plasma standard solution, Specpure™ S 10,000μg/mL
Sulfur, plasma standard solution, Specpure™ S 10,000μg/mL
Sulfur, plasma standard solution, Specpure™ S 10,000μg/mL
Thermo Scientific Chemicals

Sulfur, plasma standard solution, Specpure™ S 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014436.AE
also known as 014436-AE
Price (USD)
126.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
126.00
Each
Specifications
Chemical Name or MaterialSulfur plasma standard solution
MDL NumberMFCD00085316
Name NoteS 10000μg/mL
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only