Tin(II) iodide, ultra dry, 99.999% (metals basis)
Tin(II) iodide, ultra dry, 99.999% (metals basis)
Tin(II) iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Tin(II) iodide, ultra dry, 99.999% (metals basis)

CAS: 10294-70-9 | I2Sn | 372.519 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 014476.06
Price (USD)
170.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
170.00
Each
Chemical Identifiers
CAS10294-70-9
IUPAC Nameλ²-tin(2+) diiodide
Molecular FormulaI2Sn
InChI KeyJTDNNCYXCFHBGG-UHFFFAOYSA-L
SMILES[Sn++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Residual water< 100 ppm
Total Metal Impurities0.001% max.
Particle Size-10 Mesh
FormBeads
Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3 which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only