Yttrium(III) isopropoxide, 90+%
Yttrium(III) isopropoxide, 90+%
Yttrium(III) isopropoxide, 90+%
Yttrium(III) isopropoxide, 90+%
Thermo Scientific Chemicals

Yttrium(III) isopropoxide, 90+%

CAS: 2172-12-5 | C9H24O3Y | 269.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 014477.06
also known as 014477-06
Price (USD)
349.65
Special offer
Online exclusive
Ends: 30-Jun-2026
411.00
Save 61.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
349.65
Special offer
Online exclusive
Ends: 30-Jun-2026
411.00
Save 61.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2172-12-5
IUPAC Nametris(propan-2-ol) yttrium
Molecular FormulaC9H24O3Y
InChI KeyNREVZTYRXVBFAQ-UHFFFAOYSA-N
SMILES[Y].CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 90.0%
Yttrium(III) isopropoxide is used in asymmetric catalysis applications. It acts as a catalyst for stereoselective conjugate addition reactions and ring-opening polymerizations. It serves as a precursor for the generation of reactive enolates by enantioselective protonation reactions and nanocomposites synthesis. Further, it is used in the preparation of flat hafnium(IV) oxide films. In addition to this, it is used to improve the thermal conductivity of polycrystalline aluminum nitride (AlN) ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only