Tris(cyclopentadienyl)yttrium(III), 99.9% (REO)
Tris(cyclopentadienyl)yttrium(III), 99.9% (REO)
Tris(cyclopentadienyl)yttrium(III), 99.9% (REO)
Thermo Scientific Chemicals

Tris(cyclopentadienyl)yttrium(III), 99.9% (REO)

CAS: 1294-07-1 | C15H15Y | 284.191 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 014478.02
also known as 014478-02
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Specifications
Assay Percent Notes(REO)
CAS1294-07-1
Chemical Name or MaterialTris(cyclopentadienyl)yttrium(III)
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECS Number215-069-4
View more
Tris(cyclopentadienyl)yttrium(III) is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only