Chemical Identifiers
CAS14768-15-1
IUPAC Namegadolinium(3+) tris(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular FormulaC33H57GdO6
InChI KeyUIVVGOMXFUOLAN-UHFFFAOYSA-N
SMILES[Gd+3].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
View more