Titanium nitride, 99.7% (metals basis)
Titanium nitride, 99.7% (metals basis)
Titanium nitride, 99.7% (metals basis)
Thermo Scientific Chemicals

Titanium nitride, 99.7% (metals basis)

CAS: 25583-20-4 | NTi | 61.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 014510.A1
also known as 014510-A1
Price (USD)
804.65
Online Exclusive
894.00
Save 89.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
804.65
Online Exclusive
894.00
Save 89.35 (10%)
Each
Chemical Identifiers
CAS25583-20-4
IUPAC Nametitanioamine
Molecular FormulaNTi
InChI KeyNRTOMJZYCJJWKI-UHFFFAOYSA-N
SMILESN#[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
Titanium nitride is a hard ceramic material used as a coating on titanium alloys, steel, carbide and aluminum components. It finds application in jewelry and high-performance ceramic materials due to its gold colored surface and in medical devices such as scalpel blades and orthopedic bone saw blades. Further, it is used as a conductive barrier in microelectronics and electrodes in bioelectronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only