Barium isopropoxide, Thermo Scientific Chemicals
Barium isopropoxide, Thermo Scientific Chemicals
Barium isopropoxide, Thermo Scientific Chemicals
Barium isopropoxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium isopropoxide, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
014523.14
also known as 014523-14
25 g
Catalog number 014523.14
also known as 014523-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS24363-37-9
IUPAC Namebarium(2+) bis(propan-2-olate)
Molecular FormulaC6H14BaO2
InChI KeyCPUJSIVIXCTVEI-UHFFFAOYSA-N
SMILES[Ba++].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
Total Metal Impurities< 1.5%
Formsolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only