Lithium chloride, ultra dry, 99.9% (metals basis) - FAQs
View additional product information for Lithium chloride, ultra dry, 99.9% (metals basis) - FAQs (014540.18, 014540.30, 014540.09)
1 product FAQs found
What storage conditions should I use for Lithium chloride?
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.