Lithium chloride, ultra dry, 99.9% (metals basis)
Lithium chloride, ultra dry, 99.9% (metals basis)
Lithium chloride, ultra dry, 99.9% (metals basis)
Thermo Scientific Chemicals

Lithium chloride, ultra dry, 99.9% (metals basis)

CAS: 7447-41-8 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 014540.30
also known as 014540-30
Price (USD)
1,038.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,038.00
Each
Chemical Identifiers
CAS7447-41-8
IUPAC Namelithium(1+) chloride
Molecular FormulaClLi
InChI KeyKWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES[Li+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities0.1% max.
Residual water< 100 ppm
Particle Size-10 Mesh
FormBeads
Lithium chloride is useful for the production of lithium metal, and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer, and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere, and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis, and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Frequently asked questions (FAQs)

What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.