Aluminum chloride, ultra dry, 99.99% (metals basis)
Aluminum chloride, ultra dry, 99.99% (metals basis)
Aluminum chloride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum chloride, ultra dry, 99.99% (metals basis)

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 014552.14
also known as 014552-14
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS7446-70-0
IUPAC Namealuminium(3+) trichloride
Molecular FormulaAlCl3
InChI KeyVSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Appearance (Color)White
FormPowder
Residual water< 100 ppm
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only