Cesium sulfate, 99.9% (metals basis)
Cesium sulfate, 99.9% (metals basis)
Cesium sulfate, 99.9% (metals basis)
Thermo Scientific Chemicals

Cesium sulfate, 99.9% (metals basis)

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014559.14
also known as 014559-14
Price (USD)
83.65
Online Exclusive
92.70
Save 9.05 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.70
Save 9.05 (10%)
Each
Chemical Identifiers
CAS10294-54-9
IUPAC Namedicaesium(1+) sulfate
Molecular FormulaCs2O4S
InChI KeyFLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only