Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Holmium(III) acetate hydrate, REacton™, 99.9% (REO)

CAS: 312619-49-1 | C6H9HoO6 | 342.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 014591.18
also known as 014591-18
Price (USD)
354.65
Online Exclusive
394.00
Save 39.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
354.65
Online Exclusive
394.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS312619-49-1
IUPAC Nameholmium(3+) triacetate
Molecular FormulaC6H9HoO6
InChI KeyHWQRVFRWLQGBFV-UHFFFAOYSA-K
SMILES[Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Holmium(III) acetate hydrate is used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only