Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)

CAS: 29843-90-1 | Br3Nd | 383.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014606.14
also known as 014606-14
Price (USD)
45.30
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
45.30
Each
Add to cart
Chemical Identifiers
CAS29843-90-1
IUPAC Nameneodymium(3+) tribromide
Molecular FormulaBr3Nd
InChI KeyLBWLQVSRPJHLEY-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only