Praseodymium(III) acetate hydrate, 99.9% (REO)
Praseodymium(III) acetate hydrate, 99.9% (REO)
Praseodymium(III) acetate hydrate, 99.9% (REO)
Praseodymium(III) acetate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Praseodymium(III) acetate hydrate, 99.9% (REO)

CAS: 334869-74-8 | C6H9O6Pr | 318.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014611.22
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Chemical Identifiers
CAS334869-74-8
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Praseodymium Acetate is used to make color glasses and enamels; when mixed with certain other materials, Praseodymium produces an intense clean yellow color in glass. The other main application is used to manufacturing pigment and ink color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only