Barium 2,4-pentanedionate
Barium 2,4-pentanedionate
Barium 2,4-pentanedionate
Thermo Scientific Chemicals

Barium 2,4-pentanedionate

CAS: 12084-29-6 | C10H14BaO4 | 335.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014613.14
also known as 014613-14
Price (USD)
65.65
Online Exclusive
72.90
Save 7.25 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
65.65
Online Exclusive
72.90
Save 7.25 (10%)
Each
Chemical Identifiers
CAS12084-29-6
IUPAC Namebarium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14BaO4
InChI KeyDJHZYHWLGNJISM-FDGPNNRMSA-L
SMILES[Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ba 36% min
Barium 2,4-pentanedionate hydrate is used in the preparation of radioluminescence of poly ethylene glycolated Eu3+ doped nanophosphors as bioimaging probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only