Lanthanum(III) isopropoxide, La 40%
Lanthanum(III) isopropoxide, La 40%
Lanthanum(III) isopropoxide, La 40%
Thermo Scientific Chemicals

Lanthanum(III) isopropoxide, La 40%

CAS: 19446-52-7 | C9H21LaO3 | 316.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 014616.04
also known as 014616-04
Price (USD)
142.65
Special offer
Online exclusive
Ends: 30-Jun-2026
168.00
Save 25.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
142.65
Special offer
Online exclusive
Ends: 30-Jun-2026
168.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS19446-52-7
IUPAC Namelanthanum(3+) tris(propan-2-olate)
Molecular FormulaC9H21LaO3
InChI KeySORGMJIXNUWMMR-UHFFFAOYSA-N
SMILES[La+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
Assay (Titration)>/= 40% La
Lanthanum(III) isopropoxide finds application in asymmetric catalysis. It forms complex with an (S,S)-phenyl-linked-binaphthol ,which enhances a catalytic 1,4-addition of malonate to cyclic and acyclic enones. It acts as a co-catalyst with copper fluoride-triphenylphosphine in catalytic allylation of ketoimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only