Matrix oil, 75 centistoke hydrocarbon oil
Thermo Scientific Chemicals

Matrix oil, 75 centistoke hydrocarbon oil

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 pt.
1 gal.
Catalog number 014621.MQ
also known as 014621-MQ
Price (USD)
53.80
Each
Quantity:
1 pt.
Request bulk or custom format
Price (USD)
53.80
Each
Chemical Identifiers
CAS8042-47-5
SpecificationsSpecification SheetSpecification Sheet
CommentCOA includes trace metal concentrations.
CommentBlank
Viscosity75 centistoke
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only