Vanadium foil, 0.25mm (0.01in) thick, 99.5% (metals basis)
Vanadium foil, 0.25mm (0.01in) thick, 99.5% (metals basis)
Vanadium foil, 0.25mm (0.01in) thick, 99.5% (metals basis)
Thermo Scientific Chemicals

Vanadium foil, 0.25mm (0.01in) thick, 99.5% (metals basis)

Form: approx. 13g/50x150mm | CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 150 mm
150 x 200 mm
Catalog number 014633.FZ
also known as 014633-FZ
Price (USD)
195.65
Special offer
Online exclusive
Ends: 30-Jun-2026
230.00
Save 34.35 (15%)
Each
Quantity:
50 x 150 mm
Request bulk or custom format
Price (USD)
195.65
Special offer
Online exclusive
Ends: 30-Jun-2026
230.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.5% (metals basis)
FormFoil, 0.25mm (0.01in) thick.
Appearance (Color)White silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only