Gallium Indium Tin eutectic, 99.99% (metals basis)
Thermo Scientific Chemicals

Gallium Indium Tin eutectic, 99.99% (metals basis)

Ga:In:Sn; 62:22:16 wt.%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 014634.03
also known as 014634-03
Price (USD)
39.65
Online exclusive
43.40
Save 3.75
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Online exclusive
43.40
Save 3.75
Each
Specifications
Assay Percent Range99.99% (Metals basis)
Chemical Name or MaterialGallium Indium Tin eutectic
ColorWhite
DOT InformationDOT Class: 8, Packing Group: III
Density6.359
View more
Gallium-Indium eutectic (EGAIn) is an electrically conductive fluid metal. It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only