Antimony powder, -200 mesh, 99.999% (metals basis)
Antimony powder, -200 mesh, 99.999% (metals basis)
Antimony powder, -200 mesh, 99.999% (metals basis)
Thermo Scientific Chemicals

Antimony powder, -200 mesh, 99.999% (metals basis)

CAS: 7440-36-0 | Sb | 121.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 014640.03
also known as 014640-03
Price (USD)
38.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.10
Each
Chemical Identifiers
CAS7440-36-0
IUPAC Nameantimony
Molecular FormulaSb
InChI KeyWATWJIUSRGPENY-UHFFFAOYSA-N
SMILES[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Comment-200 Mesh
Appearance (Color)Gray
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only