Copper(I) chloride, 99.999% (metals basis)
Copper(I) chloride, 99.999% (metals basis)
Copper(I) chloride, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper(I) chloride, 99.999% (metals basis)

CAS: 7758-89-6 | ClCu | 98.996 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 014644.14
also known as 014644-14
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS7758-89-6
IUPAC Nameλ¹-copper(1+) chloride
Molecular FormulaClCu
InChI KeyOXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES[Cl-].[Cu+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only