Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL
Thermo Scientific Chemicals

Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014652.AE
also known as 014652-AE
Price (USD)
157.65
Special offer
Online exclusive
Ends: 30-Jun-2026
185.00
Save 27.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
157.65
Special offer
Online exclusive
Ends: 30-Jun-2026
185.00
Save 27.35 (15%)
Each
Specifications
Chemical Name or MaterialPrecious Metals, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECS Number231-595-7
Health Hazard 1Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Health Hazard 2May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage
View more
Precious Metals, plasma standard solution is useful as a standard for the determination of metals by inductively coupled plasma-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only