Calcium hydroxide, ACS, 95.0% min
Calcium hydroxide, ACS, 95.0% min
Calcium hydroxide, ACS, 95.0% min
Thermo Scientific Chemicals

Calcium hydroxide, ACS, 95.0% min

Air Sensitive | CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 014662.A1
also known as 014662-A1
Price (USD)
80.65
Online Exclusive
89.80
Save 9.15 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.80
Save 9.15 (10%)
Each
Chemical Identifiers
CAS1305-62-0
IUPAC Namecalcium dihydroxide
Molecular FormulaCaH2O2
InChI KeyAXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSulfur compounds (as SO₄) 0.1%
Assay (Aqueous acid-base Titration)95.0% CaH₂O₂ min
Heavy Metals(ICP-OES) 0.003%
Impurity contentCaCO₃ 3.0%
Impurity contentCl 0.03%
View more
Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materialsCalcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials
RUO – Research Use Only