Arsenic(V) oxide, 99.9% (metals basis)
Arsenic(V) oxide, 99.9% (metals basis)
Arsenic(V) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Arsenic(V) oxide, 99.9% (metals basis)

CAS: 1303-28-2 | As2O5 | 229.838 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014668.22
also known as 014668-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1303-28-2
IUPAC Namediarsooxidane
Molecular FormulaAs2O5
InChI KeyCOHDHYZHOPQOFD-UHFFFAOYSA-N
SMILESO=[As](=O)O[As](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Arsenic(V) oxide is used in the preparation of metal arsenates. It is used for dye and printing industries. Further, it is used in wood preservatives, colored glass and metal adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only