Germanium(IV) ethoxide, 97%
Germanium(IV) ethoxide, 97%
Germanium(IV) ethoxide, 97%
Thermo Scientific Chemicals

Germanium(IV) ethoxide, 97%

CAS: 14165-55-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 014679.14
also known as 014679-14
Price (USD)
492.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
492.00
Each
Specifications
CAS14165-55-0
Chemical Name or MaterialGermanium(IV) ethoxide
ColorColorless
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.14 g/mL
View more
It is a sol-gel intermediate for optical fibers and photosensitive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only