Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm
Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm
Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm
Thermo Scientific Chemicals

Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm

Preparation of niobium oxide and mixed metal oxides | CAS: 3236-82-6 | C10H25NbO5 | 318.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 014689.18
also known as 014689-18
Price (USD)
573.65
Online Exclusive
637.00
Save 63.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
573.65
Online Exclusive
637.00
Save 63.35 (10%)
Each
Chemical Identifiers
CAS3236-82-6
IUPAC Namepentaethoxyniobium
Molecular FormulaC10H25NbO5
InChI KeyZTILUDNICMILKJ-UHFFFAOYSA-N
SMILESCCO[Nb](OCC)(OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)yellow to orange
FormLiquid
Elemental AnalysisTa: ≤500ppm
Total Metal Impurities≤10ppm ( excluding Ta)
Preparation of niobium oxide and mixed metal oxides.Niobium(V) ethoxide is used for the sol-gel processing of materials containing niobium oxides and mixed metal oxides. It is also used to prepare ferroelectric nanomaterials consisting of oxides of potassium, silicon and niobium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only