Titanium(IV) chloride, 99.0% min, typically 99.6%
Titanium(IV) chloride, 99.0% min, typically 99.6%
Titanium(IV) chloride, 99.0% min, typically 99.6%
Thermo Scientific Chemicals

Titanium(IV) chloride, 99.0% min, typically 99.6%

For Use in Manufacture of Titanium Pigments and Metal, As a Catalyst in organic Syntheses, in Iridescent Glass | CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
3 x 1 L
Catalog number 014713.AE
also known as 014713-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS7550-45-0
IUPAC Nametitanium(4+) tetrachloride
Molecular FormulaCl4Ti
InChI KeyXJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (unspecified)≥99.0%
Assay (unspecified)Report metals analysis
FormLiquid
In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screenTitanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only