Yttrium(III) hexafluoro-2,4-pentanedionate
Yttrium(III) hexafluoro-2,4-pentanedionate
Yttrium(III) hexafluoro-2,4-pentanedionate
Thermo Scientific Chemicals

Yttrium(III) hexafluoro-2,4-pentanedionate

CAS: 18911-76-7 | C15H3F18O6Y | 710.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 014717.03
also known as 014717-03
Price (USD)
51.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.80
Each
Chemical Identifiers
CAS18911-76-7
IUPAC Nameyttrium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Molecular FormulaC15H3F18O6Y
InChI KeyAJVDSGQQIVVYRX-UHFFFAOYSA-N
SMILES[Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofAMetals purity: ≥99.9%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only