1-Chloroadamantane, 98%
1-Chloroadamantane, 98%
1-Chloroadamantane, 98%
Thermo Scientific Chemicals

1-Chloroadamantane, 98%

CAS: 935-56-8 | C10H15Cl | 170.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 015169.14
also known as 015169-14
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS935-56-8
IUPAC Name1-chloroadamantane
Molecular FormulaC10H15Cl
InChI KeyOZNXTQSXSHODFR-UHFFFAOYSA-N
SMILESClC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥97.5%
FormCrystalline powder / chunks
Appearance (Color)White to pale grey
It is used in some dry etching masks and polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only