Europium(III) oxalate hydrate, 99.9% (REO)
Europium(III) oxalate hydrate, 99.9% (REO)
Europium(III) oxalate hydrate, 99.9% (REO)
Europium(III) oxalate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Europium(III) oxalate hydrate, 99.9% (REO)

CAS: 152864-32-9 | C6Eu2O12 | 567.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 015288.04
also known as 015288-04
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.00
Save 8.35 (15%)
Each
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.00
Save 8.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS152864-32-9
IUPAC Namedieuropium(3+) trioxalate
Molecular FormulaC6Eu2O12
InChI KeyPVDYMOCCGHXJAK-UHFFFAOYSA-H
SMILES[Eu+3].[Eu+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only