Neodymium(III) carbonate hydrate, 99.9% (REO)
Neodymium(III) carbonate hydrate, 99.9% (REO)
Neodymium(III) carbonate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium(III) carbonate hydrate, 99.9% (REO)

CAS: 38245-38-4 | C3Nd2O9 | 468.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 015301.18
also known as 015301-18
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS38245-38-4
IUPAC Namedineodymium(3+) tricarbonate
Molecular FormulaC3Nd2O9
InChI KeyUTWHRPIUNFLOBE-UHFFFAOYSA-H
SMILES[Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Violet or pale purple to dark purple
FormPowder or crystalline powder or crystals
Total Rare Earth Oxide Impurities≤0.15% (≤1500ppm)
Neodymium Carbonate mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only