Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol
Thermo Scientific Chemicals

Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol

CAS: 79-41-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
3 kg
Catalog number 015546.A1
also known as 015546-A1
Price (USD)
84.20
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
84.20
Each
Chemical Identifiers
CAS79-41-4
IUPAC Name2-methylprop-2-enoic acid
Molecular FormulaC4H6O2
InChI KeyCERQOIWHTDAKMF-UHFFFAOYSA-N
SMILESCC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
StabilizerMEHQ (180 to 275 ppm)
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.0%
Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly (methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only