Oxalacetic acid, 98+%
Oxalacetic acid, 98+%
Oxalacetic acid, 98+%
Thermo Scientific Chemicals

Oxalacetic acid, 98+%

CAS: 328-42-7 | C4H4O5 | 132.071 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 015789.04
also known as 015789-04
Price (USD)
46.20
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
46.20
Each
Chemical Identifiers
CAS328-42-7
IUPAC Name2-oxobutanedioic acid
Molecular FormulaC4H4O5
InChI KeyKHPXUQMNIQBQEV-UHFFFAOYSA-N
SMILESOC(=O)CC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay from Suppliers CofA≥98.0 to ≤102.0% (Titration)
Chloride content≤0.01%
Residue on ignition≤0.1% (as sulfate)
Infrared spectrumConforms to structure
View more
Oxaloacetic acid is a substrate for malate dehydrogenase and oxaloacetate decarboxylase. It is an inhibitor of succinic dehydrogenase. It is an intermediate in the citric acid cycle and glucogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only