Phenol Red sodium salt, ACS
Phenol Red sodium salt, ACS
Phenol Red sodium salt, ACS
Thermo Scientific Chemicals

Phenol Red sodium salt, ACS

pH indicator | CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 015987.14
also known as 015987-14
Price (USD)
80.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
80.60
Each
Chemical Identifiers
CAS34487-61-1
IUPAC Namesodium 4-[3-(4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H13NaO5S
InChI KeyHKHYOKBQJILTEI-UHFFFAOYNA-M
SMILES[Na+].OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentClarity of solution P.T.
CommentVisual transition interval from pH 6.8 (yellow) to 8.2 (red)
Phenol red sodium salt is a pH indicator commonly used in cell culture for monitoring pH changes. It is used in the pH ranges between 6.8 and 8.2. It is involved in the studies of gastrointestinal and other systems. It can be used to check hydrogen peroxide in supernatants from cultured macrophages in multiwell plates. Further, it is used as a diagnostic aid for determination of renal function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only