alpha-Terpineol, 96%
alpha-Terpineol, 96%
alpha-Terpineol, 96%
Thermo Scientific Chemicals

alpha-Terpineol, 96%

A naturally-occuring monoterpene alcohol | CAS: 98-55-5 | C10H18O | 154.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 016285.14
also known as 016285-14
Price (USD)
34.20
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
34.20
Each
Chemical Identifiers
CAS98-55-5
IUPAC Name2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
Molecular FormulaC10H18O
InChI KeyWUOACPNHFRMFPN-VIFPVBQESA-N
SMILESCC1=CC[C@@H](CC1)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)95% min ( α + ϒ)
FormLiquid/solidified melt
CommentProduct may congeal in cold or room temperature. Warm and mix thoroughly to homogenize liquid before using or sampling
A naturally-occuring monoterpene alcohol.Alpha-terpineol is used as an antioxidant, antiseptic, antihypernociception and anti-inflammatory. It is also used as a solvent. It is an important ingredient of pine oil disinfectants. Further, it is used as a fragrance in perfumes, fat denaturant for soap production and synthetic flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only