Pyrazine, 98%
Pyrazine, 98%
Pyrazine, 98%
Thermo Scientific Chemicals

Pyrazine, 98%

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 016326.18
also known as 016326-18
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS290-37-9
IUPAC Namepyrazine
Molecular FormulaC4H4N2
InChI KeyKYQCOXFCLRTKLS-UHFFFAOYSA-N
SMILESC1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder or flakes
Appearance (Color)White to off-white
Assay from Supplier's CofA≥97.5%
Melting Point51.0-56.0°
Pyrazine is used as a flavoring agent. It serves as a component of some herbs in traditional Chinese medicine. It is employed as pharmaceutical intermediates, perfume intermediates and involved in the preparation of other chemicals. In addition to this, it is used in the production of pyrazine 1-oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only