Strontium chloride, anhydrous, 99.5% (metals basis)
Strontium chloride, anhydrous, 99.5% (metals basis)
Strontium chloride, anhydrous, 99.5% (metals basis)
Strontium chloride, anhydrous, 99.5% (metals basis)
Thermo Scientific Chemicals

Strontium chloride, anhydrous, 99.5% (metals basis)

CAS: 10476-85-4 | Cl2Sr
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 016790.22
also known as 016790-22
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10476-85-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities≤0.5% (5000 ppm)
Particle Size-40 Mesh
FormPowder
Strontium chloride is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only