Tri-n-hexyl phosphate, 90+%
Tri-n-hexyl phosphate, 90+%
Tri-n-hexyl phosphate, 90+%
Thermo Scientific Chemicals

Tri-n-hexyl phosphate, 90+%

CAS: 2528-39-4 | C18H39O4P | 350.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 016801.03
also known as 016801-03
Price (USD)
37.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.30
Each
Specifications
CAS2528-39-4
Chemical Name or MaterialTri-n-hexyl phosphate
ColorColorless
EINECS Number219-774-8
Formula Weight350.48
View more
Used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only