4-Bromo-1-butanol, tech., cont. varying amounts of THF
4-Bromo-1-butanol, tech., cont. varying amounts of THF
4-Bromo-1-butanol, tech., cont. varying amounts of THF
Thermo Scientific Chemicals

4-Bromo-1-butanol, tech., cont. varying amounts of THF

CAS: 33036-62-3 | C4H9BrO | 153.019 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 016805.06
also known as 016805-06
Price (USD)
198.65
Online Exclusive
221.00
Save 22.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
198.65
Online Exclusive
221.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS33036-62-3
IUPAC Name4-bromobutan-1-ol
Molecular FormulaC4H9BrO
InChI KeySIJLYRDVTMMSIP-UHFFFAOYSA-N
SMILESOCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 90.0%
Appearance (Color)Clear colorless to pale yellow
Formliquid
4-Bromo-1-butanol is used to prepare 2-(4-bromo-butoxy)-tetrahydro-pyran by reacting with 3,4-dihydro-2H-pyran in the presence of amberlyst 15 as a catalyst. It reacts with dihydropyran to prepare tetrahydropyranyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only