Amberlite™ IRN-77, ion exchange resin, nuclear grade
Thermo Scientific Chemicals

Amberlite™ IRN-77, ion exchange resin, nuclear grade

Styrene-DVB; Strongly acidic cation exchange resin | CAS: 11128-94-2 | Styrene-DVB
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 016821.36
also known as 016821-36
Price (USD)
44.65
Online exclusive
49.70
Save 5.05
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
44.65
Online exclusive
49.70
Save 5.05
Each
Chemical Identifiers
CAS11128-94-2
SpecificationsSpecification SheetSpecification Sheet
Total Metal ImpuritiesNa, Fe, Ca, Mg, Al: 50 ppm max, each ( on a dry basis)- Cu, 10 ppm, max ( on a dry basis)- Heavy metals ( as Pb), 10 ppm, max ( on a dry basis)
Moisture holding capacity48.0 - 55.0 ( Hydrogen form )
Appearance (Color)Pale brown to brown
Harmonic mean size0.600 - 0.700 mm
FormSpherical beads
View more
Amberlite™ IRN-77 is used in the primary water treatment for the removal of fission products, activated corrosion products and suspended matter. It is also used to control the pH of the reactor coolant stream by removing the excess lithium. Further, it is used in the removal of radioactive cations such as Cesium from waste streams. In addition to this, it is used as a cation exchanger to determine the sorption activity towards cesium(I) and strontium(II) using isothermal titration calorimetry and solution-depletion method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only